Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222453

Sigma-Aldrich

3-Methylvaleric acid

97% (GC)

Synonym(s):

3-Methylpentanoic acid, (±)-3-Methylvaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
116.16
Beilstein:
1720696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97% (GC)

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

196-198 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCC(C)CC(O)=O

InChI

1S/C6H12O2/c1-3-5(2)4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)

InChI key

IGIDLTISMCAULB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methylvaleric acid (3-Methylpentanoic acid) was used in the synthesis of 3-methyl-1-pentanol.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete structures of the sphingosine analog mycotoxins fumonisin B1 and AAL toxin TA : Absolute configuration of the side chains.
Shier WT, et al.
Tetrahedron Letters, 36(10), 1571-1574 (1995)
W E Mitch et al.
The Biochemical journal, 222(3), 579-586 (1984-09-15)
The effects of leucine, its metabolites, and the 2-oxo acids of valine and isoleucine on protein synthesis and degradation in incubated limb muscles of immature and adult rats were tested. Leucine stimulated protein synthesis but did not reduce proteolysis when
Yoshifumi Fujita et al.
Journal of receptor and signal transduction research, 27(4), 323-334 (2007-09-22)
Dresden G protein-coupled receptor (D-GPCR) is one of orphan G protein-coupled receptors (GPCR). Here we report the identification of the ligands and the characterization of D-GPCR. We investigated over 5000 compounds to evoke the response mediated by D-GPCR and identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service