Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

222100

Sigma-Aldrich

4-(Hexyloxy)aniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OC6H4NH2
CAS Number:
Molecular Weight:
193.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

155-158 °C/5 mmHg (lit.)

mp

43-45 °C (lit.)

SMILES string

CCCCCCOc1ccc(N)cc1

InChI

1S/C12H19NO/c1-2-3-4-5-10-14-12-8-6-11(13)7-9-12/h6-9H,2-5,10,13H2,1H3

InChI key

DJRKHTCUXRGYEU-UHFFFAOYSA-N

Application

4-(Hexyloxy)aniline was used in the synthesis of 4-hydroxy-4′-hexyloxyazobenzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Meyer et al.
Journal - Association of Official Analytical Chemists, 74(5), 766-771 (1991-09-01)
We have devised a method to quantitate the nitrosamine, 2-ethylhexyl-4-(N-methyl-N-nitrosoamino) benzoate (NPABAO), in commercial products containing the sunscreen ingredient, Padimate O. The method involves a minimum of cleanup steps to afford a nonaqueous extract from product emulsions suitable for analysis
George SM and Kim J.
Bull. Korean Chem. Soc., 30(5), 1143-1146 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service