Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222054

Sigma-Aldrich

4-Heptylbenzoyl chloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6C6H4COCl
CAS Number:
Molecular Weight:
238.75
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5218 (lit.)

density

1.002 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCCCCCCc1ccc(cc1)C(Cl)=O

InChI

1S/C14H19ClO/c1-2-3-4-5-6-7-12-8-10-13(11-9-12)14(15)16/h8-11H,2-7H2,1H3

InChI key

WHTFLTOKFXTJGV-UHFFFAOYSA-N

Application

4-Heptylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rama Mukherjee et al.
Bioorganic & medicinal chemistry letters, 14(12), 3169-3172 (2004-05-20)
New 3-O-acyl, 3-benzylidene, 3-hydrazone, 3-hydrazine, 17-carboxyacryloyl ester derivatives of betulinic acid (2-6, 8-11, 13, 17, 18, 21, and 22) were synthesized and evaluated in vitro for anti-angiogenic activity on endothelial cell cytotoxicity, specificity, and tube-formation ability. All derivatives reported here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service