Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

216941

Sigma-Aldrich

Behenic acid

99%

Synonym(s):

Docosanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20COOH
CAS Number:
Molecular Weight:
340.58
Beilstein:
1792887
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

mp

72-80 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear
hexane: soluble

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)

InChI key

UKMSUNONTOPOIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Behenic acid is a saturated fatty acid used as a chemical intermediate in the synthesis of various compounds.[1][2] Monomolecular films of stearic and behenic acid were formed on substrates 0.1M in sodium chloride, sodium bicarbonate or sodium phosphate and were investigated by IR analysis[3]

Application

Behenic acid was used to investigate the phase behavior of long-chain acids in supercritical propane[4]. It was also used in fabrication of metallic Langmuir-Blodgett (LB) films[3].

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase equilibria of long-chain carboxylic acids in supercritical Propane.
Schwarz CE, et al.
Journal of Chemical and Engineering Data, 56(4), 1116-1124 (2011)
Md Mazharul Islam et al.
Reproductive medicine and biology, 20(3), 289-298 (2021-07-16)
The present study was undertaken to clarify whether bovine sperm could take up fatty acids (FAs) and produce ATP to maintain linear motility. Frozen bovine semen was thawed in media containing either lipid mixture (LM) or FAs, and sperm motility
Observation of metallic conductivity down to 14 K in Langmuir-Blodgett films based on BEDO-TTF and behenic acid.
Ohnuki H, et al.
Physical Review. B, Condensed Matter and Materials Physics, 55(16), R10225-R10225 (1997)
Lais Alonso et al.
International journal of pharmaceutics, 434(1-2), 391-398 (2012-06-14)
Miltefosine (MT) is an alkylphospholipid approved for breast cancer metastasis and visceral leishmaniasis treatments, although the respective action mechanisms at the molecular level remain poorly understood. In this work, the interaction of miltefosine with the lipid component of stratum corneum
Mojgan Masoodi et al.
Rapid communications in mass spectrometry : RCM, 22(2), 75-83 (2007-12-07)
Bioactive lipid mediators derived from polyunsaturated fatty acids (PUFA) exhibit a range of tissue- and cell-specific activities in many physiological and pathological processes. Electrospray ionisation tandem mass spectrometry coupled to liquid chromatography (LC/ESI-MS/MS) is a sensitive, versatile analytical methodology for

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service