Skip to Content
MilliporeSigma
All Photos(2)

Documents

216631

Sigma-Aldrich

Chloromethyl phenyl sulfide

97%

Synonym(s):

(Phenylthio)methyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH2Cl
CAS Number:
Molecular Weight:
158.65
Beilstein:
1617369
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.594 (lit.)

bp

66 °C/0.2 mmHg (lit.)

density

1.184 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCSc1ccccc1

InChI

1S/C7H7ClS/c8-6-9-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

LLSMWLJPWFSMCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the high-yield thiomethylation of O-silylated lactone and ester enolates in the synthesis of α-methylene lactones and esters.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liebigs Ann. Chem., 49-49 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service