Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

214930

Sigma-Aldrich

1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2 · HCl
CAS Number:
Molecular Weight:
213.66
Beilstein:
3723332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

>300 °C (lit.)

SMILES string

Cl[H].OC(=O)C1Cc2ccccc2CN1

InChI

1S/C10H11NO2.ClH/c12-10(13)9-5-7-3-1-2-4-8(7)6-11-9;/h1-4,9,11H,5-6H2,(H,12,13);1H

InChI key

FXHCFPUEIDRTMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid was used in the synthesis of 10,10a-dihydroimidazo-[1,5-b]isoquinoline-1,3(2H,5H)-diones, inhibitor of inflammation, apoprotein B-100 biosynthesis and matrix-degrading metalloprotienase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan R Katritzky et al.
The Journal of organic chemistry, 67(23), 8224-8229 (2002-11-09)
Condensations of chiral diamines 11a-c with benzotriazole and formaldehyde gave benzotriazolyl intermediates 12a-c; similar condensations of alpha-amino-amides 10a-c with benzotriazole and paraformaldehyde gave 14a-c. Subsequent treatment of 12a-c and 14a-c with AlCl(3) led to enantiopure tricyclic 1,2,3,5,10,10a-hexahydroimidazo[1,5-b]isoquinolines 1a-c and 2,3,10,10a-tetrahydroimidazo[1,5-b]isoquinolin-1(5H)-ones
Yingjie Zhang et al.
Bioorganic & medicinal chemistry, 18(5), 1761-1772 (2010-02-23)
Histone deacetylases (HDACs) are enzymes involved in tumor genesis and development. Herein, we report a novel series of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid derivatives as HDACs inhibitors. The preliminary biological screening showed that most of our compounds exhibited potent inhibitory activity against HDACs.
R Guerrini et al.
Bioorganic & medicinal chemistry, 6(1), 57-62 (1998-03-21)
Substitution of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) in place of Gly2 in dynorphin A-(1-13)-NH2 and -(1-11)-NH2 (DYN) analogues (1 and 2) decreased the affinity to the kappa, delta, and mu receptors, and kappa selectivity. The analogue [D-Ala2, des-Gly3]DYN (4), a chimera between
Bradford H Hirth et al.
Bioorganic & medicinal chemistry letters, 15(8), 2087-2091 (2005-04-06)
A series of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid diamides that increase chloride transport in cells expressing mutant cystic fibrosis transmembrane conductance regulator (CFTR) protein has been identified from our compound library. Analoging efforts and the resulting structure-activity relationships uncovered are detailed. Compound potency
Xiaopan Zhang et al.
Bioorganic & medicinal chemistry, 19(20), 6015-6025 (2011-09-14)
A series of novel 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid derivatives were designed, synthesized and assayed for their activities against aminopeptidase N (APN/CD13) and MMP-2. The results showed that most compounds exhibited higher inhibitory activities against APN than that of MMP-2. Within this series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service