Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

212997

Sigma-Aldrich

2-Phenyl-3-butyn-2-ol

≥98%

Synonym(s):

α-Ethynyl-α-methylbenzenemethanol, 2-Hydroxy-2-phenyl-3-butyne, 2-Phenyl-2-hydroxy-3-butyne, 3-Hydroxy-3-phenyl-1-butyne, 3-Phenyl-1-butyn-3-ol, 3-Phenylbutyn-3-ol, Ethynylmethylphenylcarbinol, Methylphenylethynylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)(OH)C≡CH
CAS Number:
Molecular Weight:
146.19
Beilstein:
1100096
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

bp

102-103 °C/12 mmHg (lit.)

mp

47-49 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CC(O)(C#C)c1ccccc1

InChI

1S/C10H10O/c1-3-10(2,11)9-7-5-4-6-8-9/h1,4-8,11H,2H3

InChI key

KSLSOBUAIFEGLT-UHFFFAOYSA-N

Application

2-Phenyl-3-butyn-2-ol was used in the preparation of :
  • (3-methyl-5-isoxazolyl)(phenyl)-methanol
  • 1-(3-methyl-5-isoxazolyl)-1-phenyl-1-ethanol
  • α-methylene cyclic carbonates via reaction with CO2 catalyzed by transition metal salts in ionic liquid
  • copper (II)-promoted cycloaddtion to azides leading to triazoles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New 5-(2-ethenylsubstituted)-3 (2H)-furanones with in vitro antiproliferative activity.
Chimichi S, et al.
Tetrahedron, 59(28), 5215-5223 (2003)
Synlett, 957-957 (2006)
Yanlong Gu et al.
The Journal of organic chemistry, 69(2), 391-394 (2004-01-17)
Reactions of propargylic alcohols with CO(2) in a [BMIm][PhSO(3)]/CuCl catalytic system to produce the corresponding alpha-methylene cyclic carbonates were conducted with high yields. Mild reaction conditions, enhanced rates, improved yields, and recyclable ionic liquid catalyst systems are the remarkable features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service