Skip to Content
MilliporeSigma
All Photos(1)

Documents

211710

Sigma-Aldrich

Trimethylgermanium bromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3GeBr
CAS Number:
Molecular Weight:
197.65
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.47 (lit.)

bp

114 °C (lit.)

mp

−25 °C (lit.)

density

1.54 g/mL at 25 °C (lit.)

SMILES string

C[Ge](C)(C)Br

InChI

1S/C3H9BrGe/c1-5(2,3)4/h1-3H3

InChI key

WMFCXPWEKRAKMH-UHFFFAOYSA-N

Application

Reagent for the formation of dienolates of α,β-unsaturated esters. Also useful for methylenecyclopentane annulation reactions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Organometallic Chemistry, 373, 143-143 (1989)
The Journal of Organic Chemistry, 55, 3454-3454 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service