Skip to Content
MilliporeSigma
All Photos(2)

Documents

211591

Sigma-Aldrich

Gadolinium(III) nitrate hexahydrate

crystals and lumps, 99.9% trace metals basis

Synonym(s):

Gadolinium hexahydrate trinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
451.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

91 °C (lit.)

SMILES string

[Gd+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Gd.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

XWFVFZQEDMDSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gadolinium(III) nitrate hexahydrate is a highly water-soluble crystalline oxidizing agent widely used as a precursor for the synthesis of specialty glasses, and ceramics and as a phosphor for light-emitting applications.

Application

Gadolinium(III) nitrate hexahydrate can be used:
  • As a precursor to synthesize Gadolinium oxide (Gd2O3) nanoparticles for various applications such as photocatalysis and biomedical imaging.
  • As a dopant to prepare ZnO nanoparticles with enhanced room temperature ferromagnetism.
  • To prepare Ceria (CeO2)-based ceramics doped electrolytes for intermediate temperature solid oxide fuel cells (IT-SOFCs) .
  • As a raw material for the synthesis of Eu3+-Activated KGd2F7 red-emitting nanoparticles for white light-emitting diode.
  • In addition,it is also reported to producegreen light emission of Er3+-activated single-phased GdAlO3 phosphors for lighting applications.

Other Notes

Hydration may vary slightly

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and spectral studies of macrocyclic mono and bi nuclear Gandolium (III) complexes derived from Schiff's bases
Akilan P, et al
Indian Journal of Chemical Technology, 10, 363-366 (2003)
Growth of Gd2O3 nanoparticles inside mesoporous silica frameworks
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 168, 221-224 (2013)
Room-temperature synthesis of highly-efficient Eu3+-activated KGd2F7 red-emitting nanoparticles for white light-emitting diode
Yongqiang Zhong, et al
Nanomaterials, 12, 4397-4397 (2022)
Gadolinium doped ceria nanostructured oxide for intermediate temperature solid oxide fuel cells
S.U. Costilla-Aguilar, et al
Journal of alloys and compounds, 878, 160444-160444 (2021)
Temperature Dependence of Magnetic Susceptibility in Gadolinium Nitrate crystal
Kawashima R, Isoda H
Physica Status Solidi, 145(2), K59-K62 (1994)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service