Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

206563

Sigma-Aldrich

3-Iodobenzotrifluoride

98%

Synonym(s):

3-Iodo-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CF3
CAS Number:
Molecular Weight:
272.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.517 (lit.)

bp

82-82.5 °C/25 mmHg (lit.)

density

1.887 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cccc(I)c1

InChI

1S/C7H4F3I/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4H

InChI key

IGISPMBUGPHLBY-UHFFFAOYSA-N

General description

Sonogashira coupling reaction between 3-iodobenzotrifluoride and phenylacetylene using a palladium system has been reported.

Application

3-Iodobenzotrifluoride was used in the preparation of zinc 5,10,15,20-tetrakis(3-(trifluoromethyl)phenylethynyl)porphyrin.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael Luque et al.
Organic & biomolecular chemistry, 7(8), 1627-1632 (2009-04-04)
An efficient metal-free Sonogashira coupling protocol catalysed by DABCO is reported, where very good conversions and selectivities to the cross-coupling product were obtained under mild reaction conditions. The reported solvent-, phosphane- and metal-free protocol, that uses a cheap base as
Ming-Cheng Kuo et al.
Dalton transactions (Cambridge, England : 2003), (14)(14), 1433-1439 (2007-03-28)
Sonogashira coupling of zinc 5,10,15,20-tetraethynylporphyrin with various phenyl iodides under mild conditions afforded good yields of the corresponding zinc porphyrins. This method is applicable to a variety of aryl iodides including meso-substituted iodoporphyrin to form a conjugated star-shaped multiporphyrin. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service