Skip to Content
MilliporeSigma
All Photos(2)

Documents

203890

Sigma-Aldrich

Nickel(II) sulfate heptahydrate

99.999% trace metals basis

Synonym(s):

Nickel sulfate heptahydrate, Nickelous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 7H2O
CAS Number:
Molecular Weight:
280.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

crystals and lumps

impurities

≤15.0 ppm Trace Metal Analysis

density

1.948 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Ni++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

OGKAGKFVPCOHQW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) sulfate heptahydrate, a water-soluble solid, is widely utilized in diverse industrial applications such as electrochemistry, catalysis, electronics, and optoelectronics. Its electrochemical properties make it a valuable component in the synthesis of electrode materials, which are employed in batteries, supercapacitors, and fuel cells.

Application

Nickel(II) sulfate heptahydrate can be used in the research and development: 
  • As a vital precursor to generate nickel-rich cathode materials (NMC, NCA) for lithium-ion batteries using co-precipitation process because of its simplicity, ease of scale-up, and ability to produce a homogeneous structure at the particle scale.      
  • As a nickel source in the electrodeposition process to fabricate the Ni-NTNW (nickel nanotube network), which is then coated with the electroactive NiCo-LDH material to form the hierarchical supercapacitor electrode.  
  • As a precursor to synthesize the nickel(II) cinnamate complex, which is a key component in the preparation of the metal chelate monomers.
  • To synthesize a highly efficient oxygen evolution reaction (OER) trimetallic Fe-Co-Ni electrocatalyst with other metal sulfates .
  • To synthesize high entropy alloy for coatings to improve hardness, wear resistance and corrosion resistance.

Features and Benefits

  • High purity with trace metal analysis (=< 15 ppm) for 32 elements, suitable for batteries.   
  • High water solubility ideal for synthesizing composites for various applications.
  • Low ppm levels of metal ions including Al, K, Na, Mg, Cu, Co, etc.,7)      99.999% purity is ideal for nickel plating.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy S Kountz et al.
Journal of immunology (Baltimore, Md. : 1950), 208(10), 2390-2402 (2022-04-24)
Respiratory viruses stimulate the release of antiviral IFNs from the airway epithelium. Previous studies have shown that asthmatic patients show diminished release of type I and type III IFNs from bronchial epithelia. However, the mechanism of this suppression is not
Shuke Xiao et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(25), e2204238119-e2204238119 (2022-06-15)
The Anthropocene Epoch poses a critical challenge for organisms: they must cope with new threats at a rapid rate. These threats include toxic chemical compounds released into the environment by human activities. Here, we examine elevated concentrations of heavy metal
Timothy S Kountz et al.
Journal of immunology (Baltimore, Md. : 1950), 207(5), 1275-1287 (2021-08-15)
The airway epithelial cells (AECs) lining the conducting passageways of the lung secrete a variety of immunomodulatory factors. Among these, PGE2 limits lung inflammation and promotes bronchodilation. By contrast, IL-6 drives intense airway inflammation, remodeling, and fibrosis. The signaling that
Emre Karakus et al.
Scientific reports, 10(1), 7248-7248 (2020-05-01)
SLC10A7 represents an orphan member of the Solute Carrier Family SLC10. Recently, mutations in the human SLC10A7 gene were associated with skeletal dysplasia, amelogenesis imperfecta, and decreased bone mineral density. However, the exact molecular function of SLC10A7 and the mechanisms
Natali L Chanaday et al.
Neuron, 109(8), 1314-1332 (2021-03-13)
Store-operated calcium entry (SOCE) is activated by depletion of Ca2+ from the endoplasmic reticulum (ER) and mediated by stromal interaction molecule (STIM) proteins. Here, we show that in rat and mouse hippocampal neurons, acute ER Ca2+ depletion increases presynaptic Ca2+

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service