Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

200174

Sigma-Aldrich

2-Bromo-6-methoxynaphthalene

97%

Synonym(s):

2-Bromo-6-methoxynaphthalene, 2-Methoxy-6-bromonaphthalene, 6-Bromo-2-methoxynaphthalene, 6-Methoxy-2-bromonaphthalene, 6-Methoxynaphth-2-yl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC10H6OCH3
CAS Number:
Molecular Weight:
237.09
Beilstein:
2043874
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

106-109 °C (lit.)

functional group

bromo

SMILES string

COc1ccc2cc(Br)ccc2c1

InChI

1S/C11H9BrO/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7H,1H3

InChI key

AYFJBMBVXWNYLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Bromo-6-methoxynaphthalene was used in the synthesis of nabumetone [4-(6-methoxy-2-naphthalenyl)-2-butanone] by Heck reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Au?Ag?Pd trimetallic nanoparticles and their application as catalysts.
Tsai SH, et al.
J. Mater. Sci., 13(5), 978-980 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service