Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

195022

Sigma-Aldrich

2-Ethyl-2-phenylmalonamide monohydrate

99%

Synonym(s):

PEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(C2H5)(CONH2)2·H2O
CAS Number:
Molecular Weight:
224.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

120 °C (dec.) (lit.)

functional group

amide
phenyl

SMILES string

O.CCC(C(N)=O)(C(N)=O)c1ccccc1

InChI

1S/C11H14N2O2.H2O/c1-2-11(9(12)14,10(13)15)8-6-4-3-5-7-8;/h3-7H,2H2,1H3,(H2,12,14)(H2,13,15);1H2

InChI key

FZSBWHZDTUPYRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethyl-2-phenylmalonamide monohydrate was used in quantification of 2-ethyl-2-phenylmalonamide in serum of epilepsy patients by gas liquid chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Haidukewych et al.
Clinical chemistry, 26(11), 1537-1539 (1980-10-01)
We describe a procedure for determining 2-ethyl-2-phenylmalonamide (I) in serum of epilepsy patients dosed with primidone (II) for seizure control, by extracting the sample with chloroform under basic conditions, with use of an internal standard, 2-ethyl-2-(p-tolyl)malonamide, and without derivative formation.
Jane Bryant et al.
Biomaterials, 35(31), 8887-8894 (2014-07-30)
Human islet cell transplantation is a promising treatment for type 1 diabetes; however, long-term donor-specific tolerance to islet allografts remains a clinically unmet goal. We have previously shown that recipient infusions of apoptotic donor splenocytes chemically treated with 1-ethyl-3-(3'-dimethylaminopropyl)-carbodiimide (donor ECDI-SP)
Ludmila Mut et al.
Biomedical chromatography : BMC, 29(6), 935-952 (2014-11-18)
An automated multi-analyte screening method for the identification and quantification of 92 drugs and metabolites based on on-line solid-phase extraction-high-performance liquid chromatography-diode array detection technique was developed and successfully validated. In addition, a database with 870 entries including UV-spectra, relative/retention
Eike Müller et al.
Biomaterials, 53, 709-715 (2015-04-22)
Deciphering exogenous cues that determine stem cell fate decisions is a persisting challenge of cell biology and bioengineering. In an effort to unravel the role of spatial constraints in the cell-instructive characteristics of bone marrow microenvironments, murine hematopoietic stem and
Lionel Tortolano et al.
Journal of biomedical materials research. Part A, 103(12), 3843-3851 (2015-06-27)
The aim of this study is to investigate in vitro interactions between hydrophobic acrylate intraocular lenses (IOLs) and their biological environment. The influence of lens chemical composition and aging on fibronectin (FN) adsorption and on IOLs cytotoxicity on human lens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service