Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

190683

Sigma-Aldrich

3-Amino-5-methylthio-1H-1,2,4-triazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N4S
CAS Number:
Molecular Weight:
130.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

130-133 °C (lit.)

SMILES string

CSc1nc(N)n[nH]1

InChI

1S/C3H6N4S/c1-8-3-5-2(4)6-7-3/h1H3,(H3,4,5,6,7)

InChI key

XGWWZKBCQLBJNH-UHFFFAOYSA-N

General description

3-Amino-5-methylthio-1H-1,2,4-triazole on condensation with chloro-, bromo- and nitro-substituted 2-hydroxybenzaldehyde yields triazole derived Schiff base ligands. It is an efficient electrolyte additive in dye sensitized solar cell.

Application

3-Amino-5-methylthio-1H-1,2,4-triazole was used in the synthesis of heterocyclic disperse azo dyes based on 8-hydroxyquinoline.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahid H Chohan et al.
Journal of enzyme inhibition and medicinal chemistry, 28(5), 944-953 (2012-07-19)
A new series of four biologically active triazole derived Schiff base ligands (L(1)-L(4)) and their cobalt(II), nickel(II), copper(II) and zinc(II) complexes (1-16) have been synthesized and characterized. The ligands were prepared by the condensation reaction of 3-amino-5-methylthio-1H-1,2,4-triazole with chloro-, bromo-
Synthesis and spectroscopic properties of new hetarylazo 8-hydroxyquinolines from some heterocyclic amines.
Saylam A, et al.
Dyes and Pigments, 76(2), 470-476 (2008)
A Density Functional Theory Study of Additives in Electrolytes of a Dye Sensitized Solar Cell.
Lee M-E, et al.
Bull. Korean Chem. Soc., 34(8), 2491-2491 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service