Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

190632

Sigma-Aldrich

2-Chlorophenylacetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2CO2H
CAS Number:
Molecular Weight:
170.59
Beilstein:
1101523
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

92-95 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)Cc1ccccc1Cl

InChI

1S/C8H7ClO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

IUJAAIZKRJJZGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acidity constant (pKa) of 2-chlorophenylacetic acid in water has been evaluated.

Application

2-Chlorophenylacetic acid has been used in the synthesis of phenylacetoxy cellulosics and its halogenated derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prediction of acidity constant for substituted acetic acids in water using artificial neural networks.
Habibi-Yangjeh A and Danandeh-Jenagharad M.
Indian J. Chem. B, 46(3), 478-478 (2007)
Qizhou Dai et al.
Biomacromolecules, 5(1), 74-80 (2004-01-13)
A series of phenylacetoxy cellulosics with degrees of substitution (DS) between 1.4 and 3.0 and different halogenation (2-chloro, 3-chloro, 4-chloro, 2,4-dichloro, 3,4-dichloro, and 4-bromo) were synthesized. All the prepared phenylacetoxy cellulosics were soluble in dimethylformamide (DMF) and DMAc. The solubility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service