Skip to Content
MilliporeSigma

187828

2-tert-Butylcyclohexanol, mixture of isomers

99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)3CC6H10OH
CAS Number:
Molecular Weight:
156.27
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
236-806-6
MDL number:
Assay:
99%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

form

solid

mp

43-46 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)C1CCCCC1O

InChI

1S/C10H20O/c1-10(2,3)8-6-4-5-7-9(8)11/h8-9,11H,4-7H2,1-3H3

InChI key

DLTWBMHADAJAAZ-UHFFFAOYSA-N

General description

2-tert-Butylcyclohexanol is the major metabolite of (+/-)-2-tert-butylcyclohexanone.

Application

2-tert-Butylcyclohexanol was used in the synthesis of 2-tert-butylcyclohexyl methacrylate via reaction with methacryloyl chloride in the presence of triethylamine.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library