Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184055

Sigma-Aldrich

2-Methoxythiophenol

97%

Synonym(s):

2-Mercaptoanisole, 2-Methoxybenzenethiol, Thioguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4SH
CAS Number:
Molecular Weight:
140.20
Beilstein:
2042178
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.591 (lit.)

bp

99 °C/8 mmHg (lit.)

density

1.152 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1S

InChI

1S/C7H8OS/c1-8-6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

DSCJETUEDFKYGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxythiophenol was used in the synthesis of:
  • (4S)-N-[(2S)-2-methy-3-(2-methoxyphenylthio)propanoyl]-4-phenyloxazolidin-2-one
  • 2-(4-fluorophenyl)-6-(2-methoxyphenylsulfanyl)imidazo[1,2-a]pyridine
  • 2-(2-methoxyphenylsulfanyl)benzoic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric conjugate addition of thiols to chiral methacryloyloxazolidinones.
Tseng T-C and Wu M-J.
Tetrahedron Asymmetry, 6(7), 1633-1640 (1995)
A general and efficient method for the copper-catalyzed cross-coupling of amides and thiophenols with 6-halogenoimidazo [1, 2-a] pyridines.
Enguehard-Gueiffier C, et al.
Tetrahedron, 62(25), 6042-6049 (2006)
2-(2-Methoxyphenylsulfanyl) benzoic acid.
Li A-J, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(3), o1176-o1177 (2006)
Acute toxicity of thioguaiacol and of versalide in rodents.
K R Butterworth et al.
Food and cosmetics toxicology, 19(6), 753-755 (1981-12-01)
Jennifer A Jacobsen et al.
Journal of medicinal chemistry, 54(2), 591-602 (2010-12-30)
Fragment-based lead design (FBLD) has been used to identify new metal-binding groups for metalloenzyme inhibitors. When screened at 1 mM, a chelator fragment library (CFL-1.1) of 96 compounds produced hit rates ranging from 29% to 43% for five matrix metalloproteases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service