Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

182850

Sigma-Aldrich

Poly(styrene-co-acrylonitrile)

average Mw ~165,000 by GPC, acrylonitrile 25 wt. %, pellets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH(CN)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

mol wt

average Mw ~165,000 by GPC

composition

acrylonitrile, 25 wt. %

density

1.08 g/mL at 25 °C (lit.)

SMILES string

C=CC#N.C=Cc1ccccc1

InChI

1S/C8H8.C3H3N/c1-2-8-6-4-3-5-7-8;1-2-3-4/h2-7H,1H2;2H,1H2

InChI key

SCUZVMOVTVSBLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hygienic chemical studies on plastics (II). Residual acrylonitrile in ABS resin or AS resin and its migration into water (author's transl)].
T Tatsuno et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (97)(97), 93-97 (1979-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service