Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

178713

Sigma-Aldrich

9-Borabicyclo[3.3.1]nonane dimer

Synonym(s):

9-BBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30B2
CAS Number:
Molecular Weight:
244.03
Beilstein:
605509
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: reductant

mp

150-152 °C (lit.)

SMILES string

C1C[C@H]2CCC[C@@H](C1)B2B3[C@H]4CCC[C@@H]3CCC4

InChI

1S/C16H28B2/c1-5-13-7-2-8-14(6-1)17(13)18-15-9-3-10-16(18)12-4-11-15/h13-16H,1-12H2/t13-,14+,15-,16+

InChI key

IYDIZBOKVLHCQZ-GEEKYZPCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Air-stable, crystalline reagent used to reductively cleave cyclic acetals and ketals to monobenzylated 1,2-diols.
Employed in the hydroboration of alkynes, nitriles, and ketones.
Reagent found to reduce peroxo esters to alcohols without concomitant reduction of the peroxo linkage.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hong-Xia Jin et al.
The Journal of organic chemistry, 70(11), 4240-4247 (2005-05-21)
Reduction of organic molecules that contain a peroxy bond is broadly considered as a "risky" and uncertain operation when cleavage of the peroxy linkage is not desired. For this reason, such reduction steps are normally avoided at the planning stage
Synlett, 349-349 (1991)
Chemische Berichte, 126, 285-285 (1993)
Tetrahedron Letters, 32, 6239-6239 (1991)
Organic Process Research & Development, 10, 1076-1076 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service