Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177156

Sigma-Aldrich

3-Quinolinecarbonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

108-110 °C (lit.)

SMILES string

N#Cc1cnc2ccccc2c1

InChI

1S/C10H6N2/c11-6-8-5-9-3-1-2-4-10(9)12-7-8/h1-5,7H

InChI key

QZZYYBQGTSGDPP-UHFFFAOYSA-N

Application

3-Quinolinecarbonitrile was used as a template for EGFR inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diane H Boschelli
Current topics in medicinal chemistry, 8(10), 922-934 (2008-08-05)
In 2000, Wyeth reported that the 3-quinolinecarbonitrile ring system was a template for EGFR inhibitors. It soon became apparent that the group at C-4 of this core was responsible for kinase selectivity. A 4-(2,4-dichloro-5-methoxyanilino) substituent provided potent inhibitors of Src
Tamara Rodríguez-Cabo et al.
Analytical and bioanalytical chemistry, 407(20), 6159-6170 (2015-06-05)
Time-of-flight accurate mass spectrometry (TOF-MS), following a previous chromatographic (gas or liquid chromatography) separation step, is applied to the identification and structural elucidation of quinoline-like alkaloids in honey. Both electron ionization (EI) MS and positive electrospray (ESI+) MS spectra afforded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service