Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C5H8O
CAS Number:
Molecular Weight:
84.12
UNSPSC Code:
12162002
NACRES:
NA.23
PubChem Substance ID:
EC Number:
206-005-6
Beilstein/REAXYS Number:
102495
MDL number:
Quality Segment
assay
98%
form
liquid
refractive index
n20/D 1.434 (lit.)
bp
102 °C (lit.)
density
0.964 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C1CC2OC2C1
InChI
1S/C5H8O/c1-2-4-5(3-1)6-4/h4-5H,1-3H2
InChI key
GJEZBVHHZQAEDB-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
50.0 °F - closed cup
flash_point_c
10 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

