Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

172693

Sigma-Aldrich

Ethyl 4-methylbenzoate

99%

Synonym(s):

Ethyl p-toluate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2C2H5
CAS Number:
Molecular Weight:
164.20
Beilstein:
1863756
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

235 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1ccc(C)cc1

InChI

1S/C10H12O2/c1-3-12-10(11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

NWPWRAWAUYIELB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4-methylbenzoate was used in the synthesis of ethyl 4-(bromomethyl)benzoate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient and selective electrophilic and free radical catalytic bromination reactions of simple aromatic compounds in the presence of reusable zeolites.
Smith K, et al.
Journal of the Chemical Society. Perkin Transactions 1, 16, 2745-2752 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service