Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

157031

Sigma-Aldrich

3-Amino-4-methoxybenzenesulfonic acid

98%

Synonym(s):

4-Methoxymetanilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(OCH3)SO3H
CAS Number:
Molecular Weight:
203.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

305 °C (dec.) (lit.)

SMILES string

COc1ccc(cc1N)S(O)(=O)=O

InChI

1S/C7H9NO4S/c1-12-7-3-2-5(4-6(7)8)13(9,10)11/h2-4H,8H2,1H3,(H,9,10,11)

InChI key

FLIOATBXVNLPLK-UHFFFAOYSA-N

General description

3-Amino-4-methoxybenzenesulfonic acid in solid state exists in the zwitterionic form, 3-ammonio-4-methoxybenzenesulfonate. 3-Amino-4-methoxybenzenesulfonic acid was incorporated via direct coprecipitation between the sheets of a layered double hydroxide (LDH) of intralamellar composition Cu2Cr(OH)6.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ polymerization of aniline sulfonic acid derivatives into LDH interlamellar space probed by ESR and electrochemical studies.
Moujahid EM, et al.
Chemistry of Materials, 17(2), 2005-2005 (2005)
3-Ammonio-4-methoxybenzenesulfonate.
Gao S, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(3), o539-o540 (2005)
Pradeep D Uchil et al.
Cell host & microbe, 25(1), 87-100 (2019-01-01)
Lymph- and blood-borne retroviruses exploit CD169/Siglec-1-mediated capture by subcapsular sinus and marginal zone metallophilic macrophages for trans-infection of permissive lymphocytes. However, the impact of CD169-mediated virus capture on retrovirus dissemination and pathogenesis in vivo is unknown. In a murine model of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service