Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15390

Sigma-Aldrich

Boc-Ser(Bzl)-OH

≥99.0% (T)

Synonym(s):

N-(tert-Butoxycarbonyl)-O-benzyl-L-serine, Boc-O-benzyl-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
295.33
Beilstein:
3064461
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (T)

form

solid

optical activity

[α]20/D +20±1°, c = 2% in ethanol: water (4:1)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

58-60 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](COCc1ccccc1)C(O)=O

InChI

1S/C15H21NO5/c1-15(2,3)21-14(19)16-12(13(17)18)10-20-9-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,16,19)(H,17,18)/t12-/m0/s1

InChI key

DMBKPDOAQVGTST-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang-Han Chen et al.
Analytica chimica acta, 865, 53-59 (2015-03-04)
Polydimethylsiloxane (PDMS) is widely used for microfabrication and bioanalysis; however, its surface functionalization is limited due to the lack of active functional groups and incompatibility with many solvents. We presented a novel approach for in situ fabrication of cleavable peptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service