Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

153737

Sigma-Aldrich

Ethyl 4-oxo-1-piperidinecarboxylate

98%

Synonym(s):

1-Carbethoxy-4-piperidone, 1-Ethoxycarbonyl-4-piperidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
Beilstein:
640967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.475 (lit.)

density

1.135 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCOC(=O)N1CCC(=O)CC1

InChI

1S/C8H13NO3/c1-2-12-8(11)9-5-3-7(10)4-6-9/h2-6H2,1H3

InChI key

LUBGFMZTGFXIIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4-oxo-1-piperidinecarboxylate (1-carbethoxy-4-piperidone) was used to prepare tertiary alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Gundersen et al.
Bioorganic & medicinal chemistry letters, 15(7), 1891-1894 (2005-03-23)
Alzheimer's disease (AD) is a debilitating disease widely thought to be associated with the accumulation of beta amyloid (Abeta) in the brain. Inhibition of gamma-secretase, one of the enzymes responsible for Abeta production, may be a useful strategy for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service