Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14654

Sigma-Aldrich

1-Butyl-3-methylpyridinium bis(trifluormethylsulfonyl)imide

≥97.0% (H-NMR)

Synonym(s):

BMPIm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16F6N2O4S2
CAS Number:
Molecular Weight:
430.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (H-NMR)

form

liquid

impurities

<0.5% water

functional group

fluoro

SMILES string

CCCC[n+]1cccc(C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C10H16N.C2F6NO4S2/c1-3-4-7-11-8-5-6-10(2)9-11;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-6,8-9H,3-4,7H2,1-2H3;/q+1;-1

InChI key

NOFBAVDIGCEKOQ-UHFFFAOYSA-N

Application

1-Butyl-3-methylpyridinium bis(trifluormethylsulfonyl)imide can be used:
  • In the modification of electrodes applicable in the detection of dopamine and uric acid in the human urine sample.
  • As a solvent in the Ullmann homocoupling of aryl halides or aryl boronic acids using Cu nanoparticles.
  • To modify the carbon paste electrode applicable in the detection of dopamine in human serum and urine samples.

Physical form

fluorated, hydrophobic ionic liquid

Legal Information

Product protected by U.S. Patent 5,827,602 assigned to Covalent Associates, Inc. Woburn, MA, USA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the usefulness of a novel electrochemical sensor in detecting uric acid and dopamine in the presence of ascorbic acid using a screen-printed carbon electrode modified with single walled carbon nanotubes and ionic liquids
Nagles E, et al.
Electrochimica Acta, 258, 512-523 (2017)
An effective and environment-friendly system for Cu NPs@ RGO-catalyzed C-C homocoupling of aryl halides or arylboronic acids in ionic liquids under microwave irradiation
Zhao H, et al.
Royal Society of Chemistry Advances, 6(47), 41108-41113 (2016)
Imidazolium-based ionic liquids as modifiers of carbon paste electrodes for trace-level voltammetric determination of dopamine in pharmaceutical preparations
vSekuljica S, et al.
Journal of Molecular Liquids, 306, 112900-112900 (2020)

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service