Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

144487

Sigma-Aldrich

2-Amino-3-benzyloxypyridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O
CAS Number:
Molecular Weight:
200.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

92-94 °C (lit.)

functional group

phenyl

SMILES string

Nc1ncccc1OCc2ccccc2

InChI

1S/C12H12N2O/c13-12-11(7-4-8-14-12)15-9-10-5-2-1-3-6-10/h1-8H,9H2,(H2,13,14)

InChI key

NMCBWICNRJLKKM-UHFFFAOYSA-N

Gene Information

human ... MAPK14(1432)

General description

2-Amino-3-benzyloxypyridine on condensation with diethyl ethoxymethylene malonate affords 9-benzyloxy-3-ethoxycarbonylpyrido[1,2-a]pyrimidin-4-one.

Application

2-Amino-3-benzyloxypyridine was used in the synthesis of 1-acetyl-2-[2-(3-benzyloxypyridinyl)]iminoimidazolidine.

Biochem/physiol Actions

2-Amino-3-benzyloxypyridine is an inhibitor of mitogen-activated protein kinase p38α activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Chang-Fong et al.
Chemical & pharmaceutical bulletin, 48(5), 729-733 (2000-05-24)
In order to obtain possible veinotonic drugs acting through alpha2 receptor activation, we prepared clonidine analogues in which the 2-imino-imidazolidine was attached to various aliphatic or aromatic heterocycles. Among them, the two benzopyranic derivatives 16 and 22 exhibited interesting affinities
Jung-Eun Park et al.
European journal of medicinal chemistry, 141, 322-334 (2017-10-17)
Herein, we report synthesis and evaluation of new twenty six small molecules against β amyloid (Aβ)-induced opening of mitochondrial permeability transition pore (mPTP) using JC-1 assay which measures the change of mitochondrial membrane potential (ΔΨm). The neuroprotective effect of seventeen
Synthesis of new heterocyclic phenols: 9-Hydroxypyrido [1, 2-a] pyrimidin-4-one and Derivatives.
Dennin F, et al.
Journal of Heterocyclic Chemistry, 28(5), 1287-1291 (1991)
Ahmed Elkamhawy et al.
European journal of medicinal chemistry, 128, 56-69 (2017-02-06)
Herein, we report a new series of aliphatic substituted pyridyl-urea small molecules synthesized as potential modulators for amyloid beta (Aβ) induced mitochondrial dysfunction. Their blocking activities against Aβ-induced mitochondrial permeability transition pore (mPTP) opening were evaluated by JC-1 assay which
Yuya Kodama et al.
Journal of medicinal chemistry, 56(22), 9342-9350 (2013-11-01)
In this study, we developed an assignment-free approach for rapid identification of ligand-binding sites in target proteins by using NMR. With a sophisticated cell-free stable isotope-labeling procedure that introduces (15)N- or (13)C-labels to specific atoms of target proteins, this approach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service