Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

143820

Sigma-Aldrich

2,4,4-Trimethyl-2-pentene

99%

Synonym(s):

β-Diisobutylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH=C(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1719492
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

83 mmHg ( 37.7 °C)

Assay

99%

form

liquid

autoignition temp.

586 °F

refractive index

n20/D 1.416 (lit.)

bp

104 °C (lit.)

mp

−106.3-−106 °C (lit.)

density

0.72 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\C(C)(C)C

InChI

1S/C8H16/c1-7(2)6-8(3,4)5/h6H,1-5H3

InChI key

LAAVYEUJEMRIGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Butene oligomerization over phosphoric acid: structural characterization of products.
Bekker R and Prinsloo NM.
Industrial & Engineering Chemistry Research, 48(22), 10156-10162 (2009)
Kinetics of liquid-phase hydrogenation of isooctenes on a Pd/?-alumina catalyst.
Sarkar A, et al.
AIChE Journal, 52(3), 1142-1156 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service