Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139459

Sigma-Aldrich

Methyl crotonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOOCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
1720292
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

118-120 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

COC(=O)\C=C\C

InChI

1S/C5H8O2/c1-3-4-5(6)7-2/h3-4H,1-2H3/b4-3+

InChI key

MCVVUJPXSBQTRZ-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl crotonate undergoes vinylogous aldol reaction with enolizable aldehydes in the presence of aluminum tris(2,6-di-2-naphthylphenoxide).

Application

Methyl crotonate was used to investigate chemoselectivity in the reaction between methyl crotonate and benzylamine catalyzed by lipase B from Candida antarctica using solvent engineering. It was used as starting reagent during the total synthesis of phytotoxins solanapyrones D(1) and E(2).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hisahiro Hagiwara et al.
The Journal of organic chemistry, 67(17), 5969-5976 (2002-08-17)
The phytotoxins solanapyrones D (1) and E (2) have been synthesized from the decalone prepared by the domino Michael reaction of the kinetic enolate of optically pure acetylcyclohexene with methyl crotonate. The decalone was transformed into a solanapyrone core by
Solvent engineering: an effective tool to direct chemoselectivity in a lipase-catalyzed Michael addition.
Priego J, et al.
Tetrahedron, 65(2), 536-539 (2009)
Jeffrey A Gazaille et al.
Organic letters, 14(11), 2678-2681 (2012-05-25)
The synthesis of the novel Lewis acid, aluminum tris(2,6-di-2-naphthylphenoxide) (ATNP), and its use in the vinylogous aldol reaction between methyl crotonate and enolizable aldehydes are described. ATNP is related to Yamamoto's Lewis acid, aluminum tris(2,6-diphenylphenoxide) (ATPH), but the 2-naphthyl groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service