Skip to Content
MilliporeSigma
All Photos(1)

Documents

136107

Sigma-Aldrich

5-Aminoisoquinoline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

125-128 °C (lit.)

SMILES string

Nc1cccc2cnccc12

InChI

1S/C9H8N2/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H,10H2

InChI key

DTVYNUOOZIKEEX-UHFFFAOYSA-N

General description

5-Aminoisoquinoline forms 1:1 host−guest inclusion complex with β-cyclodextrin. It enhances the chemiluminescence of luminol-H2O2-horseradish peroxidase.

Application

5-Aminoisoquinoline (5AIQ) was used to study the effect of addition of β-cyclodextrin on the absorption and emission properties of 5AIQ.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Host-guest complexation between 5-aminoisoquinoline and ?-cyclodextrin and its effect on spectral and prototropic characteristics.
Rajamohan R, et al.
Journal of Inclusion Phenomena and Macrocyclic Chemistry, 73(1-4), 99-108 (2012)
Relation between the structure of some heterocyclic derivatives and other compounds, and their effects as enhancers or inhibitors of the luminol-H2O2-horseradish peroxidase chemiluminescence.
Garcia SF, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 105(!), 11-14 (1997)
Jürgen Bosch et al.
Journal of medicinal chemistry, 49(20), 5939-5946 (2006-09-29)
The 1.8 A resolution de novo structure of nucleoside 2-deoxyribosyltransferase (EC 2.4.2.6) from Trypanosoma brucei (TbNDRT) has been determined by SADa phasing in an unliganded state and several ligand-bound states. This enzyme is important in the salvage pathway of nucleoside
Sheikh Fayaz Ahmad et al.
Molecular immunology, 63(2), 394-405 (2014-10-12)
Increasing indication is unveiling a role for poly(ADP-ribose) polymerase (PARP)-1 in the regulation of inflammatory/immune responses. The aim of the present study was to determine the potential anti-inflammatory effects of PARP-1 inhibitor 5-aminoisoquinolinone (5-AIQ) to explore the role of PARP-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service