Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12803

Sigma-Aldrich

Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate

technical, ≥90% (NT)

Synonym(s):

3-(Benzothiazol-2-ylthio)-1-propanesulfonic acid sodium salt, ZPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10NNaO3S3
CAS Number:
Molecular Weight:
311.38
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical

Assay

≥90% (NT)

impurities

≤10% 3-hydroxypropanesulfonic acid sodium salt

mp

≥320 °C (dec.)

functional group

sulfonic acid
thioether

SMILES string

[Na+].[O-]S(=O)(=O)CCCSc1nc2ccccc2s1

InChI

1S/C10H11NO3S3.Na/c12-17(13,14)7-3-6-15-10-11-8-4-1-2-5-9(8)16-10;/h1-2,4-5H,3,6-7H2,(H,12,13,14);/q;+1/p-1

InChI key

VRKNGZAPJYUNSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate may be used in the chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service