Skip to Content
MilliporeSigma
All Photos(2)

Documents

127272

Sigma-Aldrich

1-Adamantaneacetic acid

98%

Synonym(s):

1-Adamantylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O2
CAS Number:
Molecular Weight:
194.27
Beilstein:
641412
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chunks

mp

134-137 °C (lit.)

SMILES string

OC(=O)CC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C12H18O2/c13-11(14)7-12-4-8-1-9(5-12)3-10(2-8)6-12/h8-10H,1-7H2,(H,13,14)/t8-,9+,10-,12-

InChI key

AOTQGWFNFTVXNQ-GOCCLTDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Adamantaneacetic acid was used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Biochem/physiol Actions

1-Adamantaneacetic acid is an inhibitor of chorismate mutase-prephenate dehydrogenase (EC 1.3.1.12) from Escherichia coli K12 (strain JP 232).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua E Mealy et al.
Advanced materials (Deerfield Beach, Fla.), 30(20), e1705912-e1705912 (2018-03-31)
Injectable hydrogels are useful for numerous biomedical applications, such as to introduce therapeutics into tissues or for 3D printing. To expand the complexity of available injectable hydrogels, shear-thinning and self-healing granular hydrogels are developed from microgels that interact via guest-host
G D Smith et al.
The Biochemical journal, 165(1), 121-126 (1977-07-01)
Several derivatives of phenylalanine and tyrosine were prepared and tested for inhibition of chorismate mutase-prephenate dehydrogenase (EC 1.3.1.12) from Escherichia coli K12 (strain JP 232). The best inhibitors were N-toluene-p-sulphonyl-L-phenylalanine, N-benzenesulphonyl-L-phenylalanine and N-benzloxycarbonyl-L-phenylalanine. Consequently two compounds, N-toluene-sulphonyl-L-p-aminophenylalanine and N-p-aminobenzenesulphonyl-L-phenylalanine, were
Alexander Trifonov et al.
ACS nano, 13(8), 8630-8638 (2019-07-17)
Through a careful chemical and bioelectronic design we have created a system that uses self-assembly of enzyme-nanoparticle hybrids to yield bioelectrocatalytic functionality and to enable the harnessing of electrical power from biomass. Here we show that mixed populations of hybrids
Xue Li et al.
ChemMedChem, 15(3), 274-283 (2019-11-26)
Nanomedicine recently emerged as a novel strategy to improve the performance of radiotherapy. Herein we report the first application of radioenhancers made of nanoscale metal-organic frameworks (nanoMOFs), loaded with gemcitabine monophosphate (Gem-MP), a radiosensitizing anticancer drug. Iron trimesate nanoMOFs possess
Małgorzata Sleszyńska et al.
International journal of peptide research and therapeutics, 18(2), 117-124 (2012-05-18)
In the current work we present some pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) modified in the N-terminal part of the molecule with a variety of acyl substituents. Of the many acylating agents used previously with B(2) receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service