Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

127256

Sigma-Aldrich

N,N-Diethyltrimethylsilylamine

97%

Synonym(s):

(Diethylamino)trimethylsilane, N-(Trimethylsilyl)diethylamine, TMSDEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NSi(CH3)3
CAS Number:
Molecular Weight:
145.32
Beilstein:
635718
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

125-126 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Si](C)(C)C

InChI

1S/C7H19NSi/c1-6-8(7-2)9(3,4)5/h6-7H2,1-5H3

InChI key

JOOMLFKONHCLCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diethyltrimethylsilylamine is a silylating agent used to derivatize polar organic compounds.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Said Kinani et al.
Journal of chromatography. A, 1190(1-2), 307-315 (2008-04-09)
This study compares different silylation procedures of zearalenone and its metabolites: alpha-zearalenol, beta-zearalenol, zearalanone, alpha-zearalanol and beta-zearalanol for gas chromatography-mass spectrometry (GC-MS) analysis. Four silylating agents among the most frequently used to derivatize polar organic compounds were tested: N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA)
Sujatha Chilakala et al.
Analytical biochemistry, 566, 62-66 (2018-11-13)
A method is proposed in this paper for the determination of oxygen-18 labeled phosphate so that positional isotope experiments using sensitive and rapid liquid chromatography-QTOF-mass spectrometry (LC-QTOF-MS) experiments can be carried out. The positional isotope exchange technique is a useful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service