Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

124621

Sigma-Aldrich

Methyl 5-bromosalicylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3-2-(OH)CO2CH3
CAS Number:
Molecular Weight:
231.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

61-65 °C (lit.)

functional group

bromo

SMILES string

COC(=O)c1cc(Br)ccc1O

InChI

1S/C8H7BrO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,1H3

InChI key

FJYDBKPPGRZSOZ-UHFFFAOYSA-N

Application

Methyl 5-bromosalicylate was used as starting reagent in the synthesis of immobilization precursor of photoswitchable piperidine base.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ragnar S Stoll et al.
Organic letters, 11(21), 4790-4793 (2009-10-02)
The synthesis of a photoswitchable piperidine base, carrying a monochlorosilane anchoring group, and its immobilization on silica gel, mimicking an oxide surface, is reported. Efficient photoswitching between the E and Z isomers of the azobenzene photochrome was demonstrated for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service