Skip to Content
MilliporeSigma
All Photos(1)

Documents

124583

Sigma-Aldrich

Diethyl azelate

technical grade, 90%

Synonym(s):

Diethyl nonanedioate, Azelaic acid diethyl ester, Diethyl nonanedioate, Nonanedioic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2C(CH2)7CO2C2H5
CAS Number:
Molecular Weight:
244.33
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

Assay

90%

refractive index

n20/D 1.435 (lit.)

bp

172 °C/18 mmHg (lit.)

mp

−16-−15.8 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCCCCC(=O)OCC

InChI

1S/C13H24O4/c1-3-16-12(14)10-8-6-5-7-9-11-13(15)17-4-2/h3-11H2,1-2H3

InChI key

CQMYCPZZIPXILQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youri M van Nuland et al.
Microbial cell factories, 16(1), 185-185 (2017-11-04)
Medium chain length (C6-C12) α,ω-dicarboxylic acids (DCAs) and corresponding esters are important building blocks for the polymer industry. For DCAs of 12 carbon atoms and longer, a sustainable process based on monooxygenase catalyzed ω-oxidation of fatty-acids has been realized. For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service