Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

118389

Sigma-Aldrich

1,3-Dimethylcyclohexane, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.426 (lit.)

bp

121-124 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC(C)C1

InChI

1S/C8H16/c1-7-4-3-5-8(2)6-7/h7-8H,3-6H2,1-2H3

InChI key

SGVUHPSBDNVHKL-UHFFFAOYSA-N

General description

cis-1,3-Dimethylcyclohexane undergoes ring opening reaction via dicarbene mechanism in the presence of iridium catalyst supported on SiO2.

Application

1,3-Dimethylcyclohexane was used to study ring opening reaction of 1,3-dimethylcyclohexane over Iridium catalysts modified by the addition of K and Ni.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ring opening of 1, 3-dimethylcyclohexane on Ir catalysts: Modification of product distribution by addition of Ni and K to improve fuel properties.
Dokjampa S, et al.
J. Mol. Catal. A: Chem., 274(1), 231-240 (2007)
Ring opening of 1, 2-and 1, 3-dimethylcyclohexane on iridium catalysts.
Do PT, et al.
J. Catal., 238(2), 477-488 (2006)
Masahiro Kono et al.
Journal of natural products, 74(3), 391-394 (2011-02-15)
Retinal analogues have been used to probe the chromophore binding pocket and function of the rod visual pigment rhodopsin. Despite the high homology between rod and cone visual pigment proteins, conclusions drawn from rhodopsin studies should not necessarily be extrapolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service