Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

118192

Sigma-Aldrich

Rhodanine

97%

Synonym(s):

2-Thioxo-4-thiazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3NOS2
CAS Number:
Molecular Weight:
133.19
Beilstein:
110701
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

165-169 °C (lit.)

solubility

methanol: soluble 2.5%, clear (yellow-green to orange-brown)

SMILES string

O=C1CSC(=S)N1

InChI

1S/C3H3NOS2/c5-2-1-7-3(6)4-2/h1H2,(H,4,5,6)

InChI key

KIWUVOGUEXMXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhodanine possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Application

Rhodanine has been used in tannase assay in cultures of tannin degrading fungi.

Biochem/physiol Actions

Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil S Cutshall et al.
Bioorganic & medicinal chemistry letters, 15(14), 3374-3379 (2005-06-18)
Dual-specificity phosphatases (DSPs) are a subclass within the protein tyrosine phosphatase family (PTPs). A series of rhodanine-based inhibitors was synthesized and shown to be novel, potent, and selective inhibitors against the DSP family member JNK-stimulating phosphatase-1 (JSP-1). Compounds of this
Culture conditions for the production of a tannase of Aspergillus tamarii IMI388810 (B).
Enemuor SC and Odibo FJC.
African Journal of Biotechnology, 8(11), 2554-2557 (2009)
H J Eggers et al.
Science (New York, N.Y.), 167(3916), 294-297 (1970-01-16)
A search for compounds which have previously unrecognized antiviral activity led to the discovery that rhodanine inhibits the multiplication of echovirus 12 and also the development of virus-induced morphologic changes. Eighteen derivatives and analogs of rhodanine were synthesized and tested
Ming-Xia Song et al.
European journal of medicinal chemistry, 54, 403-412 (2012-06-19)
Five series of (Z)-5-(4-(2-oxo-2-phenylethoxy)benzylidene)-2-thioxothiazolidin-4-one derivatives (I-V) were synthesized, characterized, and evaluated for their anti-bacterial activity. Most of the synthesized compounds showed potent inhibition against several Gram-positive bacteria (including multidrug-resistant clinical isolates) with MIC values in the range of 1-32 μg/mL.
Matjaž Brvar et al.
Bioorganic & medicinal chemistry, 20(8), 2572-2580 (2012-03-27)
Bacterial DNA gyrase is an established and validated target for the development of novel antibacterials. In our previous work, we identified a novel series of bacterial gyrase inhibitors from the class of 4-(2,4-dihydroxyphenyl) thiazoles. Our ongoing effort was designated to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service