Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

113034

Sigma-Aldrich

Oxalyl bromide

97%

Synonym(s):

Ethanedioyl dibromide, NSC 96957

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCOCOBr
CAS Number:
Molecular Weight:
215.83
Beilstein:
1744437
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.522 (lit.)

bp

16-17 °C/10 mmHg (lit.)

mp

−19 °C (lit.)

storage temp.

2-8°C

SMILES string

BrC(=O)C(Br)=O

InChI

1S/C2Br2O2/c3-1(5)2(4)6

InChI key

JAZLVNXWYDFQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in synthetic applications of carbon-substituted iminium salts

Reactant for:
  • Synthesis of mutasynthons added to cultures of A. pretiosum for mutasynthetic generation of ansamitocin derivatives
  • Oxalic acid formation from hydroxyl radical substitutions
  • Cyclization to produce CRF1 receptor antagonists
  • Preparation, optical and electrochemical studies of thiophene end capped olig(2,3-alkylthieno[3,4-b]pyrazine)
  • Asymmetrical synthesis of glycosyl chlorides and bromides
Converts cyclopentane-1,3-dione to 3-bromocyclopenten-2-one.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Luparia et al.
Organic letters, 8(10), 2147-2150 (2006-05-05)
[reaction: see text] We describe a new approach to 2,3-disubstituted cyclopentenols and cyclopentenones through two consecutive regioselective additions of equal or different electrophiles to a cyclopentene bisanionic synthon. Indeed, on exposure to BuLi, 3-bromo-2-iodocyclopent-2-enol O-TBS ether undergoes iodine-lithium permutation with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service