Skip to Content
MilliporeSigma
All Photos(2)

Documents

112984

Sigma-Aldrich

2,5-Dimethoxyaniline

98%

Synonym(s):

Aminohydroquinone dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3NH2
CAS Number:
Molecular Weight:
153.18
Beilstein:
776823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

78-80 °C (lit.)

SMILES string

COc1ccc(OC)c(N)c1

InChI

1S/C8H11NO2/c1-10-6-3-4-8(11-2)7(9)5-6/h3-5H,9H2,1-2H3

InChI key

NAZDVUBIEPVUKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dimethoxyaniline is used as a laccase inducer which immobilizes the enzyme laccase from Trumetes versicolor.
2,5-Dimethoxyaniline is used for the electrochemical copolymerization of diphenylamine in 4M sulfuric acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang CH, et al.
Journal of the Electrochemical Society, 153(5), E85-E93 (2006)
Louis Sieuw et al.
Scientific reports, 7(1), 4847-4847 (2017-07-09)
The electrochemistry of poly(2,5-dihydroxyaniline) (PDHA), a novel hybrid molecular configuration with redox active sites and electrical charge conduction along the polymer chain, has been recently reported. The theoretical capacity of this material is estimated at 443 mAh g
C Crecchio et al.
Biotechnology and bioengineering, 48(6), 585-591 (1995-12-20)
Gelatine gels originate from water in oil microemulsions in which the ternary system consists of isooctane/ sulfosuccinic acid bis [2-ethyl hexyl] ester/water; the solubilization of gelatin in the water pool of these microemulsions transforms them into viscous gels in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service