Skip to Content
MilliporeSigma
All Photos(1)

Documents

112771

Sigma-Aldrich

Methyl isothiocyanate

97%

Synonym(s):

Isothiocyanatomethane, Isothiocyanic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NCS
CAS Number:
Molecular Weight:
73.12
Beilstein:
605319
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

21 mmHg ( 20 °C)

Quality Level

Assay

97%

bp

117-118 °C (lit.)

mp

30-34 °C (lit.)

solubility

H2O: slightly soluble

density

1.069 g/cm3 at 25 °C (lit.)

SMILES string

CN=C=S

InChI

1S/C2H3NS/c1-3-2-4/h1H3

InChI key

LGDSHSYDSCRFAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl isothiocyanate (MITC) is a common soil fumigant byproduct that is toxic and corrosive, skin and respiratory irritant. It can be degraded and removed from contaminated waters by free radical reactions†.

Methyl isothiocyanate is an active chemical used as an anti-fermentative agent in wine and as an effective insecticide.

Application

Methyl isothiocyanate can be used:
  • For amino acid sequence analysis
  • To study the effect of pesticide Metam (methyl isothiocyanate is its active ingredient) in the streamside microbial communities

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bio-inspired ultrasensitive colorimetric detection of methyl isothiocyanate on nylon-6 nanofibrous membrane: A comparison of biological thiol reactivities
Peixin T et al.
Journal of Hazardous Materials, 362, 375-382 (2019)
Microbial ecology, toxicology and chemical fate of methyl isothiocyanate in riparian soils from the upper Sacramento river.
Environmental Toxicology and Chemistry / Setac, 15(10), 1694-1701 (1996)
Y Zhang et al.
Journal of environmental quality, 34(5), 1566-1572 (2005-08-11)
Recent studies have observed enhanced degradation of methyl isothiocyanate (MITC) from repeated fumigation in agricultural soils. Little is known about fumigant degradation in forest and nursery soils. This study was conducted to determine degradation rates of MITC and chloropicrin (CP)
Luis O Ruzo
Pest management science, 62(2), 99-113 (2005-11-26)
Production and use of methyl bromide, a soil fumigant, are being restricted because of this chemical's deleterious effects on stratospheric ozone concentrations. Several products, some of which are currently used as soil fumigants, are being considered as possible replacements for
Michael L Dourson et al.
Regulatory toxicology and pharmacology : RTP, 58(2), 181-188 (2010-04-22)
The preplant fumigants, metam-sodium, metam-potassium, and dazomet undergo decomposition to the biocide methyl isothiocyanate (MITC) in moist soils. Since MITC vapor can migrate from its site of application, we developed an estimate of health protective concentrations for airborne exposures to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service