Skip to Content
MilliporeSigma
All Photos(1)

Documents

112402

Sigma-Aldrich

Diethyl succinate

ReagentPlus®, 99%

Synonym(s):

1,4-Diethyl butanedioate, Diethyl butanedioate, Ethyl succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
Beilstein:
907645
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39022839
PubChem Substance ID:
NACRES:
NA.23

vapor density

6 (vs air)

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.42 (lit.)

bp

218 °C (lit.)

mp

−20 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCC(=O)OCC

InChI

1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3

InChI key

DKMROQRQHGEIOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl succinate(DES) is a diethyl ester with succinate molecules. It has two ester groups and is majorly used in fragrances. It produced by the esterification of succinic acid with ethanol.

Application

DES and 1-octanol can be blended with B5 palm oil biodiesel to improve the oxygen content and achieve a greener emission of combustion gases. It may also be used as a novel and highly efficient solvent to capture carbon dioxide(CO2) which can be potentially used as a technique to reduce carbon emission.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance and emission characteristics of green diesel blends containing diethyl-succinate and 1-octanol.
Phoon LY, et al.
Journal of Cleaner Production, 161, 1192-1202 (2017)
Diethyl succinate synthesis by reactive distillation
Orjuela A, et al.
Separation and Purification Technology, 88, 151-162 (2012)
Performance evaluation of CO2 capture with diethyl succinate.
Li H, et al.
Applied Energy, 200, 119-131 (2017)
Niki M Zacharias et al.
Journal of the American Chemical Society, 134(2), 934-943 (2011-12-08)
The Krebs tricarboxylic acid cycle (TCA) is central to metabolic energy production and is known to be altered in many disease states. Real-time molecular imaging of the TCA cycle in vivo will be important in understanding the metabolic basis of
M B Ashour et al.
Biochemical pharmacology, 36(12), 1869-1879 (1987-06-15)
A series of substituted trifluoroketones were tested as inhibitors of mammalian liver microsomal carboxylesterase(s) hydrolyzing a variety of substrates including malathion, diethylsuccinate (DES) and p-nitrophenyl acetate (p-NpAc). The trifluoroketones used were very potent "transition state" inhibitors of crude mouse and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service