Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

109398

Sigma-Aldrich

2-Norbornanemethanol, mixture of endo and exo

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.49 (lit.)

bp

93-95 °C/14 mmHg (lit.)

density

0.942 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1C[C@@H]2CC[C@H]1C2

InChI

1S/C8H14O/c9-5-8-4-6-1-2-7(8)3-6/h6-9H,1-5H2/t6-,7+,8?/m1/s1

InChI key

LWHKUVOYICRGGR-KVARREAHSA-N

General description

2-Norbornanemethanol undergoes condensation reaction with methacryloyl chloride to yield bicyclo[2.2.1]hept-2-ylmethyl 2-methylacrylate comonomer for synthesis of fluorinated poly(maleimide-co-glycidylmethacrylate).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymaleimide Copolymers with Norbornane units for Polymeric Optical Waveguides.
Kim WL, et al.
Journal of Photoscience, 9(1), 23-28 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service