Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

107212

Sigma-Aldrich

Soy Peptone for Biotechnology and Microbiology

Papain Digested, Granulated, Prime, for Biotechnology and Microbiology

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(F) · 4H2O
CAS Number:
Molecular Weight:
165.20
Beilstein:
3916184
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Product Name

Tetramethylammonium fluoride tetrahydrate, 98%

Assay

98%

form

solid

mp

39-42 °C (lit.)

functional group

amine

SMILES string

O.O.O.O.[F-].C[N+](C)(C)C

InChI

1S/C4H12N.FH.4H2O/c1-5(2,3)4;;;;;/h1-4H3;1H;4*1H2/q+1;;;;;/p-1

InChI key

HQFTZNVQVRRDLN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Homocoupling of aryl halides via a palladium catalyzed mechanism
  • Cross-coupling reactions with alkenylsilanols
  • Fluorination of dichloronitrobenzene

Used as:
  • Reversible absorbing agent for large quantities of hydrogen sulfide in the liquid state

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service