Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10720

Sigma-Aldrich

tert-Butyl 2-aminobenzoate

≥97.0%

Synonym(s):

tert-Butyl anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2
CAS Number:
Molecular Weight:
193.24
Beilstein:
2936796
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.540

density

1.06 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)c1ccccc1N

InChI

1S/C11H15NO2/c1-11(2,3)14-10(13)8-6-4-5-7-9(8)12/h4-7H,12H2,1-3H3

InChI key

HDXMPGOKFMBDHJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service