Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

105023

Sigma-Aldrich

Bromomaleic anhydride

97%

Synonym(s):

3-Bromo-2,5-dihydrofuran-2,5-dione, 3-Bromo-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HBrO3
CAS Number:
Molecular Weight:
176.95
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

refractive index

n20/D 1.54 (lit.)

bp

215 °C (lit.)

density

1.905 g/mL at 25 °C (lit.)

SMILES string

BrC1=CC(=O)OC1=O

InChI

1S/C4HBrO3/c5-2-1-3(6)8-4(2)7/h1H

InChI key

YPRMWCKXOZFJGF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service