Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

101664

Sigma-Aldrich

4-Chloro-2-nitroaniline

99%

Synonym(s):

1-Amino-4-chloro-2-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NH2
CAS Number:
Molecular Weight:
172.57
Beilstein:
512436
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

autoignition temp.

964 °F

mp

117-119 °C (lit.)

functional group

chloro
nitro

SMILES string

Nc1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C6H5ClN2O2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H,8H2

InChI key

PBGKNXWGYQPUJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

375.8 °F

Flash Point(C)

191 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D Cottalasso et al.
La Medicina del lavoro, 82(3), 253-260 (1991-05-01)
The toxicity of 4-chloro-2-nitroaniline (4C2NA) and 2-chloro-4-nitroaniline (2C4NA) was investigated on isolated rat hepatocytes following 1-3 hours of exposure to 0.2 and/or 2 mM of these xenobiotics. The higher of the two concentrations appeared to induce a statistically significant loss
H M Chopade et al.
Journal of toxicology and environmental health, 12(2-3), 267-282 (1983-08-01)
The disposition and metabolism of 14C-labeled 4-chloro-2-nitroaniline (CNA) was studied in male F344 rats following oral or intravenous (iv) administration. The gastrointestinal absorption of CNA was found to be near complete and was not affected by the dose in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service