Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

100358

Sigma-Aldrich

3-Aminoquinuclidine dihydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2 · 2HCl
CAS Number:
Molecular Weight:
199.12
Beilstein:
3681298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

321-323 °C (dec.) (lit.)

SMILES string

Cl.Cl.NC1CN2CCC1CC2

InChI

1S/C7H14N2.2ClH/c8-7-5-9-3-1-6(7)2-4-9;;/h6-7H,1-5,8H2;2*1H

InChI key

STZHBULOYDCZET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomeric purity determination of 3-aminoquinuclidine by diastereomeric derivatization and high-performance liquid chromatographic separation.
I Demian et al.
Journal of chromatography, 466, 415-420 (1989-04-19)
Christopher J Magnus et al.
Science (New York, N.Y.), 333(6047), 1292-1296 (2011-09-03)
Ionic flux mediates essential physiological and behavioral functions in defined cell populations. Cell type-specific activators of diverse ionic conductances are needed for probing these effects. We combined chemistry and protein engineering to enable the systematic creation of a toolbox of
Sofia M Morozova et al.
Membranes, 10(9) (2020-09-24)
The growing concern for climate change and global warming has given rise to investigations in various research fields, including one particular area dedicated to the creation of solid sorbents for efficient CO2 capture. In this work, a new family of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service