Merck
Discovery of adamantyl heterocyclic ketones as potent 11β-hydroxysteroid dehydrogenase type 1 inhibitors.
ChemMedChem (2011-05-25)
Xiangdong Su, Nigel Vicker, Mark P Thomas, Fabienne Pradaux-Caggiano, Heather Halem, Michael D Culler, Barry V L Potter
RESUMEN
11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) plays a key role in converting intracellular cortisone to physiologically active cortisol, which is implicated in the development of several phenotypes of metabolic syndrome. Inhibition of 11β-HSD1 activity with selective inhibitors has beneficial effects on various conditions, including diabetes, dyslipidemia and obesity, and therefore constitutes a promising strategy to discover novel therapies for metabolic and cardiovascular diseases. A series of novel adamantyl heterocyclic ketones provides potent and selective inhibitors of human 11β-HSD1. Lead compounds display low nanomolar inhibition against human and mouse 11β-HSD1 and are selective with no activity against 11β-HSD2 and 17β-HSD1. Selected potent 11β-HSD1 inhibitors show moderate metabolic stability upon incubation with human liver microsomes and weak inhibition of human CYP450 enzymes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1-Adamantyl bromomethyl ketone, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad