Merck
  • N-methyl-substituted fluorescent DAG-indololactone isomers exhibit dramatic differences in membrane interactions and biological activity.
N-methyl-substituted fluorescent DAG-indololactone isomers exhibit dramatic differences in membrane interactions and biological activity.
Chembiochem : a European journal of chemical biology (2012-10-30)
Noga Gal, Sofiya Kolusheva, Noemi Kedei, Andrea Telek, Taiyabah A Naeem, Nancy E Lewin, Langston Lim, Poonam Mannan, Susan H Garfield, Saïd El Kazzouli, Dina M Sigano, Victor E Marquez, Peter M Blumberg, Raz Jelinek
RESUMEN
N-methyl-substituted diacylglycerol-indololactones (DAG-indololactones) are newly synthesized effectors of protein kinase C (PKC) isoforms and exhibit substantial selectivity between RasGRP3 and PKCα. We present a comprehensive analysis of membrane interactions and biological activities of several DAG-indololactones. Translocation and binding activity assays underline significant variations between the PKC translocation characteristics affected by the ligands as compared to their binding activities. In parallel, the fluorescent properties of the ligands were employed for analysis of their membrane association profiles. Specifically, we found that a slight change in the linkage to the indole ring resulted in significant differences in membrane binding and association of the DAG-indololactones with lipid bilayers. Our analysis shows that seemingly small structural modifications of the hydrophobic regions of these biomimetic PKC effectors contribute to pronounced modulation of membrane interactions of the ligands.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Methylindole-2-carboxaldehyde, 97%