Merck
Histological and electrophysiological analysis of the corticospinal pathway to forelimb motoneurons in common marmosets.
Neuroscience research (2015-06-21)
Takahiro Kondo, Yamato Yoshihara, Kimika Yoshino-Saito, Tomofumi Sekiguchi, Akito Kosugi, Yuta Miyazaki, Yukio Nishimura, Hirotaka J Okano, Masaya Nakamura, Hideyuki Okano, Tadashi Isa, Junichi Ushiba
RESUMEN
Using histological and electrophysiological methods, we identified the neuroanatomical properties of the common marmoset corticospinal tract (CST), which underlies hand/arm motor control. Biotinylated dextran amine (BDA) was injected into the primary motor cortex to anterogradely label CST axons in the cervical segments, revealing that most CST axons descend in the contralateral dorsolateral funiculus (DLF; 85.0%), and some in the ipsilateral DLF (10.7%). Terminal buttons were mainly found in the contralateral lamina VII of the gray matter, but projection to lamina IX, where forelimb motoneurons are located, was rare. Bilateral projections were more abundant than found in the rat CST, resembling the CST organization of other primates. Intracellular recordings were made from 57 forelimb motoneurons on the contralateral side to stimulation, which revealed no monosynaptic excitatory postsynaptic potentials (EPSPs), but di- or polysynaptic EPSPs and inhibitory synaptic potentials were commonly found. Local field potentials showed monosynaptic excitation mainly in laminae VII, where abundant BDA-labeled CST terminals were observed. These results suggest that direct corticomotoneuronal projection is absent in common marmosets but di- or oligosynaptic effects would be mediated by spinal interneurons.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, for molecular biology, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, BioUltra, for molecular biology, ≥99.5% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, Grade I, ≥99% (GC), suitable for plant cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, meets USP testing specifications
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, ACS reagent
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización